Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012771.1 | XP_016868260.1 | 13 | 2755 | ||
GenBank mRNA | DQ218280.1 | - | 1 | 807 | ||
DQ218281.1 | - | 1 | 1175 |