Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL6P28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037979.1 | - | 1 | 755 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI522604.1 | - | 1 | 755 | Chr. 22 - 44901547 - 44902458 [+] on Build GRCh38 |