Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027827.1 | XP_016883316.1 | 12 | 2405 | ||
GenBank mRNA | AK095985.1 | - | 1 | 230 |