Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UROS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC024936.1 | - | 1 | 315 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG028484.1 | - | 1 | 314 | Chr. 10 - 125791845 - 125792571 [-] on Build GRCh38 |