Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263496.4 | XP_005263553.1 | 4 | 1886 | ||
XM_005263497.4 | XP_005263554.1 | 3 | 1730 | |||
GenBank mRNA | AK026883.1 | - | 1 | 815 |