Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCIRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057093.1 | - | 1 | 1301 |
Gene Symbol: CHKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU617910.1 | - | 1 | 91 |