Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308028.1 | NP_001294957.1 | 18 | 7539 | ||
NM_001308031.1 | NP_001294960.1 | 11 | 6489 | |||
NM_005246.3 | NP_005237.2 | 20 | 7925 | |||
XM_011543276.2 | XP_011541578.1 | 12 | 6741 | |||
GenBank mRNA | AK092320.1 | - | 1 | 293 | ||
AK123847.1 | - | 1 | 1154 |