Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025349.1 | XP_016880838.1 | 6 | 784 | ||
XM_017025350.1 | XP_016880839.1 | 6 | 784 | |||
XM_017025351.1 | XP_016880840.1 | 6 | 784 | |||
XM_017025356.1 | XP_016880845.1 | 5 | 866 | |||
GenBank mRNA | AK023469.1 | - | 1 | 333 | ||
AK296120.1 | - | 4 | 755 | |||
AK302523.1 | - | 5 | 865 |