Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039614.2 | NP_001034703.1 | 2 | 3885 | ||
NM_001303254.1 | NP_001290183.1 | 3 | 3768 | |||
XM_005254369.4 | XP_005254426.1 | 3 | 5168 | |||
GenBank mRNA | AK124841.1 | - | 1 | 1269 | ||
AL360257.1 | - | 1 | 19 |