Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511395.2 | XP_011509697.1 | 35 | 10770 | ||
GenBank mRNA | AL117611.1 | - | 1 | 844 |