Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IREB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL117613.1 | - | 762 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB164928.1 | - | 1 | 307 | Chr. 4 - 159335025 - 159335623 [+] on Build GRCh38 |