Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF319524.1 | - | LIT2203 IC-SNURF-SNRPN 3 prime noncoding transcripts |
9 | 1627 | |
AF400485.1 | - | LIT2216 IC-SNURF-SNRPN 3 prime noncoding transcripts |
3 | 309 |