Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268157.2 | XP_005268214.1 | 10 | 2305 | ||
XM_005268159.2 | XP_005268216.1 | 9 | 2134 | |||
XM_005268160.4 | XP_005268217.1 | 7 | 3401 | |||
XM_011537276.1 | XP_011535578.1 | 10 | 1715 | |||
XM_017021729.1 | XP_016877218.1 | 10 | 2197 | |||
XM_017021730.1 | XP_016877219.1 | 9 | 2026 | |||
XM_017021731.1 | XP_016877220.1 | 9 | 2035 | |||
XM_017021732.1 | XP_016877221.1 | 9 | 4364 | |||
GenBank mRNA | AK093741.1 | - | 1 | 2177 |