Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021732.1 | XP_016877221.1 | 9 | 2370 | ||
GenBank mRNA | AK093741.1 | - | 1 | 183 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB108808.1 | - | 1 | 183 | Chr. 14 - 50331103 - 50331667 [-] on Build GRCh38 |