Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251313.2 | XP_005251370.1 | 9 | 6581 | ||
XM_017013883.1 | XP_016869372.1 | 9 | 6681 | |||
XM_017013885.1 | XP_016869374.1 | 9 | 6389 | |||
GenBank mRNA | AK022971.1 | - | 1 | 937 |