Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLRN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126442.1 | - | 1, 2 | 127 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB270441.1 | - | 1, 2 | 127 | Chr. 3 - 150852484 - 151079072 [+] on Build GRCh38 |