Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM47E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242936.1 | NP_001229865.1 | 4 | 778 | ||
GenBank mRNA | AK124936.1 | - | 4 | 778 | ||
AK310457.1 | - | 3 | 404 | |||
BC020802.1 | - | 1 | 1297 | |||
DB081518.1 | - | 3 | 404 |
Gene Symbol: FAM47E-STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090914.1 | - | 2 | 175 | ||
AK296418.1 | - | 3 | 371 |