Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125525.1 | - | 1 | 1413 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA871839.1 | - | 1 | 333 | Chr. X - 15827653 - 15828197 [-] on Build GRCh38 |