Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012417.3 | NP_036549.2 | 9 | 5590 | ||
NM_181671.2 | NP_858057.1 | 10 | 5709 | |||
XM_005257216.2 | XP_005257273.1 | 9 | 5090 | |||
XM_017024442.1 | XP_016879931.1 | 9 | 4701 | |||
XM_017024443.1 | XP_016879932.1 | 10 | 4820 | |||
XM_017024444.1 | XP_016879933.1 | 10 | 5211 | |||
XM_017024445.1 | XP_016879934.1 | 10 | 4833 | |||
GenBank mRNA | AK094724.1 | - | 1 | 1539 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM690777.1 | - | 1 | 110 | Chr. 17 - 67696503 - 67697116 [+] on Build GRCh38 |