Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002564.3 | NP_002555.3 | 3 | 6121 | ||
NM_176071.2 | NP_788085.2 | 3 | 6186 | |||
NM_176072.2 | NP_788086.2 | 3 | 6255 | |||
GenBank mRNA | AK094727.1 | - | 1 | 972 |
Set Membership: |
G-protein coupled receptor |