Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005254228.3 | XP_005254285.1 | 28 | 7629 | ||
XM_011521363.2 | XP_011519665.1 | 28 | 7359 | |||
GenBank mRNA | AL117454.1 | - | 1 | 1701 |