Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017009024.1 | XP_016864513.1 | 7 | 1753 | ||
GenBank mRNA | AK124911.1 | - | 1 | 1140 | ||
AK126876.1 | - | 1 | 3790 |