Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270407.1 | NP_001257336.1 | 9 | 3472 | ||
NM_021219.3 | NP_067042.1 | 10 | 3580 | |||
NR_072999.1 | 10 | 3576 | ||||
GenBank mRNA | AK056079.1 | - | 1 | 1663 | ||
BG540412.1 | - | 1 | 332 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR423676.1 | - | 1 | 462 | Chr. 21 - 25716354 - 25717164 [+] on Build GRCh38 |