Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270407.1 | NP_001257336.1 | 9 | 3686 | ||
NM_021219.3 | NP_067042.1 | 10 | 3794 | |||
NR_072999.1 | 10 | 3790 | ||||
GenBank mRNA | AK056079.1 | - | 1 | 1877 | ||
BG540412.1 | - | 1 | 546 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR423676.1 | - | 1 | 676 | Chr. 21 - 25716354 - 25717164 [+] on Build GRCh38 |