Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258046.3 | XP_005258103.1 | 9 | 3120 | ||
XM_011525358.1 | XP_011523660.1 | 9 | 3123 | |||
XM_011525359.1 | XP_011523661.1 | 9 | 3106 | |||
XM_011525362.1 | XP_011523664.1 | 9 | 3004 | |||
XM_011525367.1 | XP_011523669.1 | 7 | 2911 | |||
XM_011525368.2 | XP_011523670.1 | 7 | 2540 | |||
XM_011525369.2 | XP_011523671.1 | 7 | 2688 | |||
XM_011525370.2 | XP_011523672.1 | 7 | 2685 | |||
XM_017025228.1 | XP_016880717.1 | 9 | 3135 | |||
XM_017025229.1 | XP_016880718.1 | 9 | 2860 | |||
XM_017025230.1 | XP_016880719.1 | 8 | 2614 | |||
GenBank mRNA | AF086453.1 | - | 1 | 67 |