Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274051.2 | XP_005274108.1 | 16 | 8167 | ||
XM_017017923.1 | XP_016873412.1 | 16 | 7915 | |||
GenBank mRNA | AK096617.1 | - | 1 | 1737 |