Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047404.1 | - | 1 | 892 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI630872.1 | - | 1 | 323 | Chr. 10 - 73253280 - 73253755 [+] on Build GRCh38 |