Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134459.1 | 2 | 654 | |||
NR_134460.1 | 3 | 686 | ||||
GenBank mRNA | BC033983.1 | - | 2 | 609 | ||
BC047404.1 | - | 1 | 1481 |