Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251731.3 | XP_005251788.1 | 1 | 942 | ||
GenBank mRNA | AK057300.1 | - | 1 | 942 |