Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282116.1 | NP_001269045.1 | 17 | 7355 | ||
NM_134428.2 | NP_602304.1 | 18 | 7422 | |||
XM_006716840.1 | XP_006716903.1 | 18 | 7331 | |||
XM_006716843.1 | XP_006716906.1 | 19 | 7331 | |||
XM_006716844.1 | XP_006716907.1 | 19 | 7347 | |||
XM_006716847.3 | XP_006716910.1 | 13 | 6464 | |||
XM_011517998.1 | XP_011516300.1 | 19 | 7399 | |||
XM_011517999.1 | XP_011516301.1 | 19 | 7363 | |||
XM_011518000.1 | XP_011516302.1 | 17 | 7334 | |||
XM_011518001.1 | XP_011516303.1 | 20 | 7413 | |||
XM_017015000.1 | XP_016870489.1 | 19 | 7350 | |||
XM_017015001.1 | XP_016870490.1 | 15 | 7025 | |||
XM_017015002.1 | XP_016870491.1 | 12 | 8870 | |||
GenBank mRNA | AK127804.1 | - | 1 | 2808 |