Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525004.1 | XP_011523306.1 | 1 | 1363 | ||
XM_011525006.1 | XP_011523308.1 | 1 | 1363 | |||
XM_017024850.1 | XP_016880339.1 | 1 | 1363 | |||
XM_017024851.1 | XP_016880340.1 | 1 | 1363 | |||
XM_017024852.1 | XP_016880341.1 | 1 | 1363 | |||
XM_017024853.1 | XP_016880342.1 | 1 | 1363 | |||
XM_017024854.1 | XP_016880343.1 | 1 | 1363 | |||
GenBank mRNA | AK025174.1 | - | 1 | 224 | ||
BX647700.1 | - | 2 | 2719 |