Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271754.1 | NP_001258683.1 | 11 | 3298 | ||
XM_005254100.4 | XP_005254157.1 | 12 | 3474 | |||
GenBank mRNA | AK026784.1 | - | 1 | 316 | ||
CD110646.1 | - | 1 | 201 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG696705.1 | - | 1 | 53 | Chr. 13 - 101719220 - 101719874 [+] on Build GRCh38 |