Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290046.1 | NP_001276975.1 | 19 | 8943 | ||
NM_001290047.1 | NP_001276976.1 | 19 | 9415 | |||
XM_006724079.3 | XP_006724142.1 | 7 | 7858 | |||
XM_011546128.2 | XP_011544430.1 | 18 | 9019 | |||
XM_011546129.2 | XP_011544431.1 | 19 | 9019 | |||
XM_011546132.2 | XP_011544434.1 | 19 | 8938 | |||
XM_017028785.1 | XP_016884274.1 | 19 | 9100 | |||
GenBank mRNA | AK055696.1 | - | 1 | 890 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ950712.1 | - | 1 | 73 | Chr. 22 - 17557370 - 17558142 [+] on Build GRCh38 |