Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023578.1 | - | 1637 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU852929.1 | - | 1 | 631 | Chr. 13 - 60572855 - 60573579 [+] on Build GRCh38 |