Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303037.1 | NP_001289966.1 | 12 | 3541 | ||
NM_016447.3 | NP_057531.2 | 13 | 3710 | |||
XM_005249775.4 | XP_005249832.1 | 11 | 3322 | |||
XM_006715738.3 | XP_006715801.1 | 12 | 3841 | |||
XM_006715739.3 | XP_006715802.1 | 12 | 3432 | |||
XM_006715740.3 | XP_006715803.1 | 12 | 3429 | |||
XM_011515425.2 | XP_011513727.1 | 12 | 3537 | |||
XM_017012315.1 | XP_016867804.1 | 12 | 3465 | |||
XM_017012316.1 | XP_016867805.1 | 12 | 3586 | |||
XM_017012317.1 | XP_016867806.1 | 12 | 3466 | |||
GenBank mRNA | BC047330.1 | - | 1 | 796 |