Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123376.2 | NP_001116848.1 | 5 | 2788 | ||
XM_011540069.2 | XP_011538371.1 | 5 | 5590 | |||
XM_011540070.2 | XP_011538372.1 | 5 | 5536 | |||
XM_011540071.2 | XP_011538373.1 | 6 | 2897 | |||
GenBank mRNA | AB235418.1 | - | 5 | 2743 | ||
BC029135.1 | - | 1 | 592 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI762258.1 | - | 592 | Chr. - - [-] on Build |