Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001249.3 | NP_001240.1 | 16 | 5146 | ||
NM_001321985.1 | NP_001308914.1 | 17 | 5263 | |||
NM_001321986.1 | NP_001308915.1 | 17 | 5243 | |||
NM_001321987.1 | NP_001308916.1 | 18 | 5390 | |||
NM_001321988.1 | NP_001308917.1 | 16 | 5922 | |||
XM_005268224.3 | XP_005268281.1 | 16 | 5108 | |||
XM_006720326.2 | XP_006720389.1 | 15 | 5008 | |||
XM_017021813.1 | XP_016877302.1 | 17 | 5268 | |||
XM_017021816.1 | XP_016877305.1 | 13 | 4882 | |||
GenBank mRNA | AK025209.1 | - | 1 | 1676 |