Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120450.1 | 1 | 663 | |||
GenBank mRNA | BC050035.1 | - | 1 | 663 |