Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024654.1 | XP_016880143.1 | 1 | 594 | ||
GenBank mRNA | AF318360.1 | - | 1 | 1134 | ||
AK001561.1 | - | 1 | 595 |