Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017854.1 | - | 2 | 549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD516739.1 | - | 1 | 618 | Chr. 12 - 13196802 - 13197569 [+] on Build GRCh38 |