Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031723.3 | NP_001026893.1 | 8 | 4966 | ||
NM_001278310.1 | NP_001265239.1 | 7 | 4794 | |||
GenBank mRNA | AK095074.1 | - | 1 | 1050 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM699445.1 | - | 1 | 689 | Chr. 4 - 99897345 - 99898045 [-] on Build GRCh38 |