Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031723.3 | NP_001026893.1 | 8 | 4322 | ||
NM_001278310.1 | NP_001265239.1 | 7 | 4150 | |||
XM_011532262.2 | XP_011530564.1 | 10 | 4206 | |||
XM_011532263.2 | XP_011530565.1 | 8 | 4706 | |||
XM_011532265.2 | XP_011530567.1 | 7 | 4650 | |||
XM_017008628.1 | XP_016864117.1 | 10 | 4388 | |||
GenBank mRNA | AK095074.1 | - | 1 | 406 |