Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UROS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016610.1 | XP_016872099.1 | 10 | 989 | ||
XM_017016611.1 | XP_016872100.1 | 10 | 945 | |||
GenBank mRNA | AK090736.1 | - | 1 | 1647 | ||
AK092076.1 | - | 1 | 2025 |