Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LETMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019151.1 | XP_016874640.1 | 8 | 1157 | ||
XM_017019152.1 | XP_016874641.1 | 8 | 1123 | |||
XM_017019153.1 | XP_016874642.1 | 7 | 1007 | |||
XM_017019154.1 | XP_016874643.1 | 7 | 970 | |||
XM_017019155.1 | XP_016874644.1 | 8 | 995 | |||
XM_017019156.1 | XP_016874645.1 | 6 | 855 | |||
XM_017019157.1 | XP_016874646.1 | 6 | 817 | |||
XM_017019158.1 | XP_016874647.1 | 7 | 858 | |||
XM_017019159.1 | XP_016874648.1 | 5 | 786 | |||
XM_017019160.1 | XP_016874649.1 | 5 | 748 | |||
XM_017019161.1 | XP_016874650.1 | 5 | 754 | |||
XM_017019162.1 | XP_016874651.1 | 5 | 724 | |||
XM_017019164.1 | XP_016874653.1 | 7 | 1024 | |||
XM_017019165.1 | XP_016874654.1 | 8 | 1086 | |||
XM_017019166.1 | XP_016874655.1 | 7 | 907 | |||
GenBank mRNA | AK123080.1 | - | 1 | 734 | ||
AK126372.1 | - | 1 | 26 |