Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104359.1 | 6 | 3086 | |||
GenBank mRNA | AK002005.1 | - | 1 | 1558 | ||
CB160399.1 | - | 1 | 367 |