Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090894.1 | - | 1 | 2843 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX501593.1 | - | 1 | 35 | Chr. 17 - 68876986 - 68877459 [-] on Build GRCh38 |