Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545388.2 | XP_011543690.1 | 1 | 616 | ||
GenBank mRNA | BC034230.1 | - | 1 | 2053 |