Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090561.1 | - | 1 | 1171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL597401.1 | - | 1 | 215 | Chr. 7 - 118187763 - 118188294 [+] on Build GRCh38 |