Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006823.3 | NP_006814.1 | 4 | 3218 | ||
NM_181839.2 | NP_862822.1 | 3 | 3089 | |||
XM_017013620.1 | XP_016869109.1 | 5 | 3093 | |||
XM_017013621.1 | XP_016869110.1 | 4 | 3059 | |||
XM_017013622.1 | XP_016869111.1 | 3 | 2824 | |||
GenBank mRNA | BX419914.2 | - | 1 | 266 |