Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720539.2 | XP_006720602.1 | 43 | 12349 | ||
XM_011521613.2 | XP_011519915.1 | 44 | 12406 | |||
XM_011521614.2 | XP_011519916.1 | 45 | 12092 | |||
XM_011521615.2 | XP_011519917.1 | 44 | 12259 | |||
XM_011521616.2 | XP_011519918.1 | 44 | 12053 | |||
XM_011521617.2 | XP_011519919.1 | 44 | 12403 | |||
XM_011521618.2 | XP_011519920.1 | 43 | 12352 | |||
XM_011521619.2 | XP_011519921.1 | 43 | 12349 | |||
XM_011521620.2 | XP_011519922.1 | 43 | 12346 | |||
XM_011521621.2 | XP_011519923.1 | 43 | 12286 | |||
XM_011521622.2 | XP_011519924.1 | 43 | 12190 | |||
XM_011521623.2 | XP_011519925.1 | 27 | 9509 | |||
XM_017022228.1 | XP_016877717.1 | 44 | 12035 | |||
XM_017022229.1 | XP_016877718.1 | 44 | 12025 | |||
XM_017022230.1 | XP_016877719.1 | 42 | 12133 | |||
GenBank mRNA | BC039397.1 | - | 1 | 1041 |